Order Entry
Poland
Orders LinkContactUsLinkComponent
Cesium fluoride, Sigma-Aldrich®

Cesium fluoride, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIAL198323-25G
Toxic
Corrosive
Supplier:   Merck
Cesium fluoride, Sigma-Aldrich®
Catalog # SIAL198323-25G
Supplier:  Merck
CAS Number:  13400-13-0
Cesium fluoride, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  13400-13-0
Price
528 zł
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:13400-13-0
  • Formula:CsF
  • MDL Number:MFCD00010960
  • Molecular Weight:151.9 g/mol
  • Boiling Pt:1251 °C (1013 hPa)
  • Melting Pt:680 °C
  • Density:4.627 g/cm³ (20 °C)
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 4,627 g/cm³ (20 °C)
  • EINECS:236-487-3
  • UN:2923
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications