Order Entry
Poland
Orders LinkContactUsLinkComponent
Selenium, Sigma-Aldrich®

Selenium, Sigma-Aldrich®

Glass bottle for solids, 20 g
Catalog # SIAL204307-20G
Toxic
Health
Supplier:   Merck
Selenium, Sigma-Aldrich®
Catalog # SIAL204307-20G
Supplier:  Merck
CAS Number:  7782-49-2
Selenium, Sigma-Aldrich®

Glass bottle for solids, 20 g

CAS Number:  7782-49-2
Price
866 zł
Log in for availability
1 * 20 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:3283
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 g

Specifications