Order Entry
Poland
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALA2218-25G
Supplier:   Merck
L(+)-Ascorbic acid, Sigma-Aldrich®
Catalog # SIALA2218-25G
Supplier:  Merck
CAS Number:  50-81-7
L(+)-Ascorbic acid, Sigma-Aldrich®
L(+)-Ascorbic acid, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  50-81-7
Price
513 zł
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.