Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Calibration oils for viscometers

1060, 475 mL
  BROOB1060
Calibration oils for viscometers
  BROOB1060
 :  
 :  
Calibration oils for viscometers
Calibration oils for viscometers

1060, 475 mL

 :  
 

Silicone fluids and hydrocarbon oils for viscometers.

  • Accurate to ±1% of viscosity value
  • NIST traceable certification

Silicone fluid standards are most commonly used for calibration of rotational viscometers. Excellent temperature stability.

Hydrocarbon oils (Mineral oils or polybutene) are appropriate for use at shear rates of greater than 500 sec-¹. Recommended for use with cone/plate viscometers at viscosities above 5000 cP.

Viscosity = nominal viscosity at 25 °C unless stated otherwise.

 

 

  • Pk:
    475 mL
  • Viscosity:
    1060