Order Entry
Poland
Orders LinkContactUsLinkComponent
Calibration oils for viscometers

Calibration oils for viscometers

29, 475 mL
Catalog # BROO21CHS29250
Calibration oils for viscometers
Catalog # BROO21CHS29250
Supplier:  
CAS Number:  
Calibration oils for viscometers
Calibration oils for viscometers

29, 475 mL

CAS Number:  
Discontinued
Product Details & Documents

Silicone fluids and hydrocarbon oils for viscometers.

  • Accurate to ±1% of viscosity value
  • NIST traceable certification

Silicone fluid standards are most commonly used for calibration of rotational viscometers. Excellent temperature stability.

Hydrocarbon oils (Mineral oils or polybutene) are appropriate for use at shear rates of greater than 500 sec-¹. Recommended for use with cone/plate viscometers at viscosities above 5000 cP.

Viscosity = nominal viscosity at 25 °C unless stated otherwise.

Documents

Specifications

Specifications

  • Pk:
    475 mL
  • Viscosity:
    29

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.