Scalpels
2, 150 mm, Scalpel, with mounted blade, yellow
Catalog # BOCH12641
Supplier: BOCHEM
Jump to:
Product Details & Documents
These mounted scalpels feature a steel blade and a coloured handle made of impact resistant PS.
- Not sterile
- Blades are exchangeable
- Spare blades available (set of 10 pieces)
Specifications
- Type:2
- Description:Scalpel, with mounted blade, yellow
- Length:150 mm