Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
POTASSIUM OXALATE MONOHYDRATE, 99%, A.C. S. REAGENT 1 * 500 g
Catalog # 223425-500G
Supplier:
Honeywell Chemicals
SDS
Expand image
POTASSIUM OXALATE MONOHYDRATE, 99%, A.C. S. REAGENT 1 * 500 g
Catalog # 223425-500G
Supplier:
Honeywell Chemicals
SDS
Related Information
Documentation
SDS
sds_223425
Documentation
Frequently Bought Together
di-Sodium L(+)-tartrate dihydrate, Sigma-Aldrich®
Supplier: Merck