Buffers, concentrated solutions, Fluka™
9,00, ±0.02, sodium tetraborate/hydrochloric acid, 500 ml, Ampoule, 1
Supplier: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Catalog # 38748-1EA
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Ampoule
- Pk:1
- pH accuracy:±0.02
- Final volume:500 ml
- pH value:9,00
- Contents:sodium tetraborate/hydrochloric acid



