Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzaldehyde, Sigma-Aldrich®
Benzaldehyde, Sigma-Aldrich®
Glass bottle, 2,5 L
  SIALB1334-2.5L
 :   Merck
Benzaldehyde, Sigma-Aldrich®
  SIALB1334-2.5L
 :  Merck
 :  100-52-7
Benzaldehyde, Sigma-Aldrich®
Benzaldehyde, Sigma-Aldrich®
Glass bottle, 2,5 L
 :  100-52-7
536 zł
1 * 2,5 L
  •  :100-52-7
  •  :C₆H₅CHO
  •  :MFCD00003299
  •  :106.12 g/mol
  •  :13,01057
  •  :179 °C (1013 hPa)
  •  :–26 °C
  •  :1.049 g/cm³ (20 °C)
  •  :64 °C
  •  :Ambient
  •  :1990
  •  :9,III
  •  :M-Clarity™ quality level = MQ200
  •  :202-860-4
Documents
  • Pack type:
    Glass bottle
  • Pk:
    2,5 L