Order Entry
Poland
Orders LinkContactUsLinkComponent
Guaiacol

Guaiacol

Glass bottle for solids, 100 g
Catalog # CAYM70430-100
Irritant
Supplier:   Cayman Chemical
Guaiacol
Catalog # CAYM70430-100
Supplier:  Cayman Chemical
CAS Number:  90-05-1
Guaiacol

Glass bottle for solids, 100 g

CAS Number:  90-05-1
Price
271 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol
  • CAS Number:90-05-1
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00002185
  • Molecular Weight:124.14 g/mol
  • Merck Index:13,04568
  • Boiling Pt:205 °C (1013 hPa)
  • Melting Pt:28…32 °C
  • Density:1.1534 g/cm³ (0 °C)
  • Flash Pt:82 °C
  • Storage Temperature:Ambient
  • EINECS:201-964-7
  • UN:2810
  • ADR:6.1,III

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications