Order Entry
Poland
Orders LinkContactUsLinkComponent
2,4-Dibromopyridine 97%

2,4-Dibromopyridine 97%

Glass bottle, 5 g
Catalog # APOSOR8632-5G
Corrosive
Toxic
Supplier:   Apollo Scientific
2,4-Dibromopyridine 97%
Catalog # APOSOR8632-5G
Supplier:  Apollo Scientific
CAS Number:  58530-53-3
2,4-Dibromopyridine 97%

Glass bottle, 5 g

CAS Number:  58530-53-3
Price
107 zł
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:58530-53-3
  • Formula:C₅H₃Br₂N
  • MDL Number:MFCD01859720
  • Molecular Weight:236.89 g/mol
  • Melting Pt:35…40 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.