Order Entry
Poland
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 100 mL
Catalog # SIALS7899-100ML
Supplier:   Merck
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®
Catalog # SIALS7899-100ML
Supplier:  Merck
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 100 mL

CAS Number:  
Price
203 zł
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)
  • Cat. No.:
    SIALS7899-100ML
  • M-Clarity quality level:
    MQ200

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.