Order Entry
Poland
Orders LinkContactUsLinkComponent
Trimethyl phosphate ≥98%

Trimethyl phosphate ≥98%

Glass bottle for solids, 2.500 g
Catalog # A10595.0E
Health
Irritant
Trimethyl phosphate ≥98%
Catalog # A10595.0E
CAS Number:  512-56-1
Trimethyl phosphate ≥98%
Trimethyl phosphate ≥98%

Glass bottle for solids, 2.500 g

CAS Number:  512-56-1
Price
1 960 zł
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 98+% CAS No.: 512-56-1 Molecular Formula: C3H9O4P Formula Weight: 140.08 Flash Point: 150°(302°F) Refractive Index: 1.3970 Odour: Pleasant Melting Point: -46° Boiling Point: 196-198° Beilstein Reference: 1071731 MDL No.: MFCD00008348

Synonyms:  Trimethylphosphate
  • CAS Number:512-56-1
  • Formula:C₃H₉O₄P
  • MDL Number:MFCD00008348
  • Molecular Weight:140.08 g/mol
  • Boiling Pt:196…198 °C
  • Melting Pt:–46 °C
  • Density:1.213
  • Flash Pt:>150 °C (302 °F)
  • Storage Temperature:Ambient
  • EINECS:208-144-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.