Order Entry
Poland
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7,5 (2 mol/L), Sigma-Aldrich®

Buffer, TRIS solution pH 7,5 (2 mol/L), Sigma-Aldrich®

TRIS buffer solution pH 7,5 (2 mol/l), Glass bottle, 1 L
Catalog # SIALT2944-1L
Supplier:   Merck
Buffer, TRIS solution pH 7,5 (2 mol/L), Sigma-Aldrich®
Catalog # SIALT2944-1L
Supplier:  Merck
CAS Number:  
Buffer, TRIS solution pH 7,5 (2 mol/L), Sigma-Aldrich®

TRIS buffer solution pH 7,5 (2 mol/l), Glass bottle, 1 L

CAS Number:  
Price
874 zł
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS buffer solution pH 7,5 (2 mol/l)
  • Cat. No.:
    SIALT2944-1L
  • M-Clarity quality level:
    MQ200

Specifications