SIGMA FAST FAST RED TR/NAPHTHOL AS MX*TA BLET SETS 1 1 * 50 items
SIALF4648-50SET
: Merck
Frequently Bought Together
n-Hexane, EMPLURA®, Supelco®
: Merck
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
: PanReac AppliChem
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
: PanReac AppliChem
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
ETER DIETYLOWY CZDA, PH.EUR. OP. 2,5 DM3 1 * 2,5 L
: POCH AVANTOR BRAND
tert-Butanol ACS
: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
METANOL DO HPLC 1 * 1 L
: CHEMPUR
Acetone 99+%, extra pure
: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
: Thermo Fisher Scientific