0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
PROTAZYME AK 1 * 200 items
MEGAT-PRAK-200T
:
MEGAZYME
PROTAZYME AK 1 * 200 items
MEGAT-PRAK-200T
:
MEGAZYME
Frequently Bought Together
EOSIN RAL 555 1L 1 * 1 L
: RAL
Test Strips, nitrate, MQuant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
VWR®, Immersion Oil
: VWR Collection
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Supelco® Spectroquant® Test Kits, Sulphate
: Merck
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck