Order Entry
Poland
Orders LinkContactUsLinkComponent
Phosphate buffer solution, pH 7,2 ( 25 °C), Fluka™

Phosphate buffer solution, pH 7,2 ( 25 °C), Fluka™

Phosphate buffer solution, 7,20, ±0,04, Plastic Bottle, 500 mL
Catalog # 319252-500ML
Supplier:   Honeywell Chemicals
Phosphate buffer solution, pH 7,2 ( 25 °C), Fluka™
Catalog # 319252-500ML
CAS Number:  
Phosphate buffer solution, pH 7,2 ( 25 °C), Fluka™

Phosphate buffer solution, 7,20, ±0,04, Plastic Bottle, 500 mL

CAS Number:  
Price
144 zł
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic Bottle
  • Pk:
    500 mL
  • Description:
    Phosphate buffer solution
  • pH accuracy:
    ±0,04
  • pH value:
    7,20

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.