Filter Paper Circles MN GF-3 4.7 cm diameter pack of 100 1 * 100 items
Catalog # MANA4130047
Supplier: MACHEREY-NAGEL
Filter Paper Circles MN GF-3 4.7 cm diameter pack of 100 1 * 100 items
Catalog # MANA4130047
Supplier: MACHEREY-NAGEL
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Weighing bottles, glass
Supplier: Lenz Laborglas GmbH & CO.KG
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® Erlenmeyer Flasks, with Standard Ground Joint
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection
Cellulose, microcrystalline ∼ Ø 50 µm
Supplier: Thermo Fisher Scientific