Order Entry
Poland
Orders LinkContactUsLinkComponent
pH Buffer solutions in bottles

pH Buffer solutions in bottles

pH Buffer solution, 7,01, ±0,01, Colourless, 25 °C, 500 mL
Catalog # HANNHI7007L
Supplier:   Hanna
pH Buffer solutions in bottles
Catalog # HANNHI7007L
Supplier:  Hanna
CAS Number:  
pH Buffer solutions in bottles

pH Buffer solution, 7,01, ±0,01, Colourless, 25 °C, 500 mL

CAS Number:  
Price
124 zł
Log in for availability
1 * 500 mL
Product Details & Documents

Plastic bottle with a tamper-proof seal.

  • Color coded label

Traceable to NIST standard reference materials.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • Description:
    pH Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    7,01

Specifications