Ribbons for IP™ label printing system
IP-R-6000, 6000 ribbon series, resin formulation, width 60 mm, 300 m, Black
BRDY066202
: Brady
These IP® printer ribbons are high density ribbon with excellent solvent resistance, good heat resistance and super scratch resistance and have ribbon spindle.
- Colour:Black
- Description:IP-R-6000, 6000 ribbon series, resin formulation, width 60 mm
- Length:300 m
- Seq:06