Order Entry
Poland
Orders LinkContactUsLinkComponent
Chromium standard solutions

Chromium standard solutions

Chromium, hexavalent, standard solution (NIST), Chromium VI, Cr, 1000 mg/L, 100 mL
Catalog # HACH1466442
Supplier:   Hach
Chromium standard solutions
Catalog # HACH1466442
Supplier:  Hach
CAS Number:  
Chromium standard solutions
Chromium standard solutions

Chromium, hexavalent, standard solution (NIST), Chromium VI, Cr, 1000 mg/L, 100 mL

CAS Number:  
Price
203 zł
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in total chromium determinations.

  • Traceable to NIST

Documents

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    100 mL
  • Description:
    Chromium, hexavalent, standard solution (NIST)
  • Element:
    Chromium VI
  • Symbol:
    Cr

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.