ANAEROB INDICATOR TEST, PGE. WITH 100 TEST STRIPS* 1 * 100 items
Catalog # SIAL59886-1PAK-F
Supplier: Merck
ANAEROB INDICATOR TEST, PGE. WITH 100 TEST STRIPS* 1 * 100 items
Catalog # SIAL59886-1PAK-F
Supplier: Merck
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Metanol CZDA, ODCZ. FP op. 1 dm3x6 1 * 1 L
Supplier: POCH AVANTOR BRAND
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl carbonate, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Water, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Butyl acetate ACS
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals