Order Entry
Poland
Orders LinkContactUsLinkComponent
Agar, Millipore®

Agar, Millipore®

Glass bottle for solids, 1 kg
Catalog # SIAL05040-1KG
Supplier:   Merck
Agar, Millipore®
Catalog # SIAL05040-1KG
Supplier:  Merck
CAS Number:  9002-18-0
Agar, Millipore®
Agar, Millipore®

Glass bottle for solids, 1 kg

CAS Number:  9002-18-0
Price
2 050 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:C₁₂nH₁₈nOn₉n
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications