VWR® Preservation System, Cryoinstant
Cryoinstant, green, 50
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.
- Quick and easy to use
- Colour coded
- Storage down to –70 °C
- Quality controlled for fertility and sterility
Packaging: Cryoinstant is packed in packs of 50 vials of 2 ml.
Certificates
Search by Lot Number822070ZA_CRYOINSTANT VIALS MIXED 50X2ML_ISO 11133_rev1 822071ZA_CRYOINSTANT VIALS RED 50X2ML_ISO 11133_rev1 822072ZA_CRYOINSTANT VIALS BLUE 50X2ML_ISO 11133_rev1 822073ZA_CRYOINSTANT VIALS GREEN 50X2ML_ISO 11133_rev1 822074ZA_CRYOINSTANT VIALS YELLOW 50X2ML_ISO 11133_rev1
More
Specifications
Specifications
- Pk:50
- Description:Cryoinstant, green
- Cat. No.:822073ZA
- Shelf life:1440



