L(-)-Proline 99+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
- CAS Number:147-85-3
- Formula:C₅H₉NO₂
- MDL Number:MFCD00064318
- Molecular Weight:115.13 g/mol
- Merck Index:13,07871
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:228…233 °C
- Density:1.35 g/cm³
- Storage Temperature:Ambient
- EINECS:205-702-2
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- UV transmittance:≥95 % (c=10, H2O, 430 nm) 1 cm cell
- pH:5.5 to 7 (2.5 % at 25°C)
- Specific optical rotation:-84.5° to -86.8° (20°C, 589 nm) (c=4, H2O)
- Titration with HClO4:99.0 to 101.0 %
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤200 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Other amino acids:≤0.5 % (TLC)
- Insoluble matter:≤0.005 % (in water)
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



