Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Benzaldehyde 98+%

Glass bottle, 250 mL
Catalog # ACRO105222500
Irritant
Benzaldehyde 98+%
Catalog # ACRO105222500
CAS Number:  100-52-7
Benzaldehyde 98+%
Benzaldehyde 98+%

Glass bottle, 250 mL

CAS Number:  100-52-7
Price
152 zł
Log in for availability
1 * 250 mL
Product Details & Documents
  • CAS Number:100-52-7
  • Formula:C₆H₅CHO
  • MDL Number:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,01057
  • Boiling Pt:179 °C (1013 hPa)
  • Melting Pt:–26 °C
  • Density:1.043 g/cm³
  • Flash Pt:64 °C
  • Storage Temperature:Ambient
  • EINECS:202-860-4
  • UN:1990
  • ADR:9,III

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5440 to 1.5460 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Specific gravity:
    (25°C) 1.041 to 1.046
  • Acid value:
    ≤5 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.