Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator
Glass bottle, 250 mL
Supplier: VWR Chemicals
Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator
Catalog # 31720.230
Supplier: VWR Chemicals
CAS Number: 547-58-0
Jump to:
Product Details & Documents
- Transition range: pH 3,1-4,4
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
- CAS Number:547-58-0
- Formula:C₁₄H₁₄N₃NaO₃S
- MDL Number:MFCD00007502
- Molecular Weight:327.34 g/mol
- Storage Temperature:Ambient
- EINECS:208-925-3
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Glass bottle
- Pk:250 mL



