5,5'-DITHIOBIS(2-NITROBENZOIC ACID) >= 98% (TLC), BIOREAGENT, SUITABLE FOR DETERMINATION OF SULFHYDRYL GROUPS 1 * 1 g
Catalog # SIALD8130-1G
Supplier: Merck
5,5'-DITHIOBIS(2-NITROBENZOIC ACID) >= 98% (TLC), BIOREAGENT, SUITABLE FOR DETERMINATION OF SULFHYDRYL GROUPS 1 * 1 g
Catalog # SIALD8130-1G
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Ascorbate oxidase (from <i>Cucumis sp.</i>)
Supplier: Merck