Pestles, serrated
235 mm, 5 ml mortars, 6,5 mm
KART6305
: KARTELL
Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.
- Head made of PTFE
- Shaft made of stainless steel
- Ø:6,5 mm
- For:5 ml mortars
- Length:235 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



