Order Entry
Poland
Orders LinkContactUsLinkComponent
L-shaped spreaders, PS | Avantor
L-shaped spreaders, PS
L-shape bacteriology loop, with special angle inclination, packs of 5
numer katalogowy KART2648
Dostawca:   KARTELL
L-shaped spreaders, PS
numer katalogowy KART2648
Dostawca:  KARTELL
L-shaped spreaders, PS
L-shaped spreaders, PS
L-shape bacteriology loop, with special angle inclination, packs of 5
Przejdź do:
Price
19,60 zł
Zaloguj się dla dostępności
1 * 5 SZT
Product Details & Documents
Disposable L-shaped sterile PS spreaders for bacteriology with special 100° angling for easier use. Especially handy for dispersing liquids across agar culture plates.
Sposób pakowania: Available single-wrapped or in packs of 5.
Dane techniczne
  • Description:
    L-shape bacteriology loop, with special angle inclination, packs of 5
  • Seq:
    01