Order Entry
Poland
Orders LinkContactUsLinkComponent
Sodium L(+)-ascorbate ≥99%

Sodium L(+)-ascorbate ≥99%

Glass bottle for solids, 500 g
Catalog # A17759.36
Sodium L(+)-ascorbate ≥99%
Catalog # A17759.36
CAS Number:  134-03-2
Sodium L(+)-ascorbate ≥99%
Sodium L(+)-ascorbate ≥99%

Glass bottle for solids, 500 g

CAS Number:  134-03-2
Price
863 zł
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 134-03-2 Molecular Formula: C6H7NaO6 Formula Weight: 198.11 Melting Point: ca 220° Beilstein Reference: 3767246 Merck Reference: 14,830 Optical Rotation: +105° (c=5 in water) MDL No.: MFCD00082340

Synonyms:  Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
  • CAS Number:134-03-2
  • Formula:C₆H₇NaO₆
  • MDL Number:MFCD00082340
  • Molecular Weight:198.11 g/mol
  • Merck Index:14,830
  • Melting Pt:∼220 °C
  • Density:1.799 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-126-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.