Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Sodium L(+)-ascorbate ≥99%

Glass bottle for solids, 100 g
Catalog # A17759.22
Sodium L(+)-ascorbate ≥99%
Catalog # A17759.22
CAS Number:  134-03-2
Sodium L(+)-ascorbate ≥99%
Sodium L(+)-ascorbate ≥99%

Glass bottle for solids, 100 g

CAS Number:  134-03-2
Price
271 zł
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 134-03-2 Molecular Formula: C6H7NaO6 Formula Weight: 198.11 Melting Point: ca 220° Beilstein Reference: 3767246 Merck Reference: 14,830 Optical Rotation: +105° (c=5 in water) MDL No.: MFCD00082340

Synonyms:  Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
  • CAS Number:134-03-2
  • Formula:C₆H₇NaO₆
  • MDL Number:MFCD00082340
  • Molecular Weight:198.11 g/mol
  • Merck Index:14,830
  • Melting Pt:∼220 °C
  • Density:1.799 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-126-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications