Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead(II) nitrate ≥99%

Lead(II) nitrate ≥99%

Glass bottle for solids, 100 g
Catalog # A16345.22
Oxidizing
Health
Irritant
Lead(II) nitrate ≥99%
Catalog # A16345.22
CAS Number:  10099-74-8
Lead(II) nitrate ≥99%
Lead(II) nitrate ≥99%

Glass bottle for solids, 100 g

CAS Number:  10099-74-8
Price
204 zł
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Odour: Odourless Melting Point: 470° UN No.: 1469 MDL No.: MFCD00011153

Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:14,5414
  • Melting Pt:470 °C
  • Density:4.530
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.