Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead(II) nitrate ≥99%

Lead(II) nitrate ≥99%

Glass bottle for solids, 2.500 g
Catalog # A16345.0E
Oxidizing
Health
Irritant
Lead(II) nitrate ≥99%
Catalog # A16345.0E
CAS Number:  10099-74-8
Lead(II) nitrate ≥99%
Lead(II) nitrate ≥99%

Glass bottle for solids, 2.500 g

CAS Number:  10099-74-8
Price
1 460 zł
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Odour: Odourless Melting Point: 470° UN No.: 1469 MDL No.: MFCD00011153

Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:14,5414
  • Melting Pt:470 °C
  • Density:4.530
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications