Order Entry
Poland
Orders LinkContactUsLinkComponent
1,8-Octanediol ≥98%
Glass bottle for solids, 100 g
  A15402.22
1,8-Octanediol ≥98%
  A15402.22
 :  629-41-4
1,8-Octanediol ≥98%
1,8-Octanediol ≥98%
Glass bottle for solids, 100 g
 :  629-41-4
811 zł
1 * 100 g
Purity: 98+% CAS No.: 629-41-4 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 148°(298°F) Melting Point: 58-61° Boiling Point: 171-173°/20mm Beilstein Reference: 1633499 MDL No.: MFCD00002989
 :  1,8-Dihydroxy octane
  •  :629-41-4
  •  :C₈H₁₈O₂
  •  :MFCD00002989
  •  :146.23 g/mol
  •  :171…173 °C (20 mmHg)
  •  :58…61 °C
  •  :0.941 g/cm³ (20 °C)
  •  :148 °C (298 °F)
  •  :Ambient
  •  :211-090-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g