Order Entry
Poland
Orders LinkContactUsLinkComponent
1,8-Octanediol ≥98%

1,8-Octanediol ≥98%

Glass bottle for solids, 25 g
Catalog # A15402.14
1,8-Octanediol ≥98%
Catalog # A15402.14
CAS Number:  629-41-4
1,8-Octanediol ≥98%
1,8-Octanediol ≥98%

Glass bottle for solids, 25 g

CAS Number:  629-41-4
Price
291 zł
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 629-41-4 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 148°(298°F) Melting Point: 58-61° Boiling Point: 171-173°/20mm Beilstein Reference: 1633499 MDL No.: MFCD00002989

Synonyms:  1,8-Dihydroxy octane
  • CAS Number:629-41-4
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00002989
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:171…173 °C (20 mmHg)
  • Melting Pt:58…61 °C
  • Density:0.941 g/cm³ (20 °C)
  • Flash Pt:148 °C (298 °F)
  • Storage Temperature:Ambient
  • EINECS:211-090-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.