Order Entry
Poland
Orders LinkContactUsLinkComponent
1,8-Octanedithiol ≥98%

1,8-Octanedithiol ≥98%

Glass bottle for solids, 25 g
Catalog # B24693.14
Irritant
1,8-Octanedithiol ≥98%
Catalog # B24693.14
CAS Number:  1191-62-4
1,8-Octanedithiol ≥98%
1,8-Octanedithiol ≥98%

Glass bottle for solids, 25 g

CAS Number:  1191-62-4
Price
1 980 zł
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 1191-62-4 Molecular Formula: HS(CH2)8SH Formula Weight: 178.36 Flash Point: 113°(235°F) Refractive Index: 1.5050 Odour: Unpleasant Boiling Point: 269-270° Beilstein Reference: 1735431 MDL No.: MFCD00003574

Synonyms:  1,8-Dimercaptooctane
  • CAS Number:1191-62-4
  • Formula:C₈H₁₈S₂
  • MDL Number:MFCD00003574
  • Molecular Weight:178.36 g/mol
  • Boiling Pt:269…270 °C
  • Density:0.960
  • Flash Pt:113 °C (235 °F)
  • Storage Temperature:Ambient
  • EINECS:214-738-8
  • UN:3334
  • ADR:9,
  •  :Density: 0.9814 g/cm³ (0 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications