1,8-Octanedithiol ≥98%
Glass bottle for solids, 5 g
Dostawca: Thermo Fisher Scientific
1,8-Octanedithiol ≥98%
numer katalogowy B24693.06
Dostawca: Thermo Fisher Scientific
Numer CAS: 1191-62-4
Przejdź do:
Product Details & Documents
Purity: 98% CAS No.: 1191-62-4 Molecular Formula: HS(CH2)8SH Formula Weight: 178.36 Flash Point: 113°(235°F) Refractive Index: 1.5050 Odour: Unpleasant Boiling Point: 269-270° Beilstein Reference: 1735431 MDL No.: MFCD00003574
Synonimy: 1,8-Dimercaptooctane
- Numer CAS:1191-62-4
- Wzór:C₈H₁₈S₂
- Numer MDL:MFCD00003574
- Molecular Weight:178.36 g/mol
- Temperatura wrzenia:269…270 °C
- Gęstość:0.960
- Punkt zapalny:113 °C (235 °F)
- Temperatura przechowywania:Ambient
- EINECS:214-738-8
- UN:3334
- ADR:9,
- :Density: 0.9814 g/cm³ (0 °C)
Documents
Dane techniczne
Dane techniczne
- Pack type:Glass bottle for solids
- Pk:5 g
Dane techniczne
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



