2-Bromoisobutyryl bromide, 97% 1 * 25 g
Catalog # B24249.14
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Cyclohexane, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) for DNA and peptides synthesis
Supplier: POCH AVANTOR BRAND
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Chemicals