Fuchsin (acid)
Glass bottle for solids, 500 g
CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286
: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
- :3244-88-0
- :C₂₀H₁₇N₃Na₂O₉S₃
- :MFCD00013286
- :585.55 g/mol
- :14,107
- :> 130 °C
- :Ambient
- :221-816-5
Documents
- Pack type:Glass bottle for solids
- Pk:500 g