3,3-Dimethyloctane ≥98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 4110-44-5 Molecular Formula: C10H22 Formula Weight: 142.29 Boiling Point: 161° UN No.: 3295 MDL No.: MFCD00048874
- CAS Number:4110-44-5
- Formula:C₁₀H₂₂
- MDL Number:MFCD00048874
- Molecular Weight:142.28 g/mol
- Boiling Pt:161 °C
- Melting Pt:–69 °C
- Density:0,74
- Storage Temperature:Temperatura otoczenia
- UN:3295
- ADR:3,III
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Rodzaj opakowania:Glass bottle for solids
- Ilość:5 g



