3,3-Dimethyloctane ≥98%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 4110-44-5 Molecular Formula: C10H22 Formula Weight: 142.29 Boiling Point: 161° UN No.: 3295 MDL No.: MFCD00048874
- CAS Number:4110-44-5
- Formula:C₁₀H₂₂
- MDL Number:MFCD00048874
- Molecular Weight:142.28 g/mol
- Boiling Pt:161 °C
- Melting Pt:–69 °C
- Density:0,74
- Storage Temperature:Temperatura otoczenia
- UN:3295
- ADR:3,III
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Rodzaj opakowania:Glass bottle for solids
- Ilość:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



