Order Entry
Poland
Orders LinkContactUsLinkComponent
N-Methylanthranilic acid ≥90%
N-Methylanthranilic acid ≥90%
Glass bottle for solids, 25 g
  B20531.14
N-Methylanthranilic acid ≥90%
  B20531.14
 :  119-68-6
N-Methylanthranilic acid ≥90%
N-Methylanthranilic acid ≥90%
Glass bottle for solids, 25 g
 :  119-68-6
259 zł
1 * 25 g
Purity: 90+% CAS No.: 119-68-6 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: ca 171° dec. Beilstein Reference: 2208135
 :  2-(Methylamino)benzoic acid
  •  :119-68-6
  •  :C₈H₉NO₂
  •  :MFCD00002424
  •  :151.16 g/mol
  •  :80 °C (0.01 mmHg)
  •  :∼171 °C (dec.)
  •  :Ambient
  •  :204-343-9
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g