Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzaldehyde ≥99%

Benzaldehyde ≥99%

Glass bottle for solids, 1.000 g
Catalog # A10348.0B
Irritant
Benzaldehyde ≥99%
Catalog # A10348.0B
CAS Number:  100-52-7
Benzaldehyde ≥99%
Benzaldehyde ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  100-52-7
Price
257 zł
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99+% CAS No.: 100-52-7 Molecular Formula: C7H6O Formula Weight: 106.12 Flash Point: 62°(143°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.5450 Sensitivity: Air Sensitive Appearance: Colourless Odour: Almond-like Melting Point: -26° Boiling Point: 178-179° Beilstein Reference: 471223 Merck Reference: 14,1058 UN No.: 1990 MDL No.: MFCD00003299

  • CAS Number:100-52-7
  • Formula:C₆H₅CHO
  • MDL Number:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:14,1058
  • Boiling Pt:178…179 °C
  • Melting Pt:–26 °C
  • Density:1.047
  • Flash Pt:62 °C (143 °F)
  • Storage Temperature:Ambient
  • EINECS:202-860-4
  • UN:1990
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications