Order Entry
Poland
ContactUsLinkComponent
Burrette titration,quick operation burettes- for rapid and accurate titration, even the smallest amounts of liquid. a gentle pressur e on the soft polyethylene storage bottle fills the glass burette. the glass burette has a schellbach stripes and an 1 * 1 items
Catalog # SEMA1259
Supplier:  SEMADENI
Burrette titration,quick operation burettes- for rapid and accurate titration, even the smallest amounts of liquid. a gentle pressur e on the soft polyethylene storage bottle fills the glass burette. the glass burette has a schellbach stripes and an 1 * 1 items
Catalog # SEMA1259
Supplier:  SEMADENI
Frequently Bought Together