Order Entry
Poland
Orders LinkContactUsLinkComponent
Analytical HPLC columns, Microsorb™ C18

Analytical HPLC columns, Microsorb™ C18

Microsorb™ MV, Analytical, C18, 100 Å, 5 µm, 4,60 mm, 150 mm
Catalog # VARIR0086200D5
Supplier:   VARIAN
Analytical HPLC columns, Microsorb™ C18
Catalog # VARIR0086200D5
Supplier:  VARIAN
Analytical HPLC columns, Microsorb™ C18

Microsorb™ MV, Analytical, C18, 100 Å, 5 µm, 4,60 mm, 150 mm

Price
2 820 zł
Log in for availability
1 * 1 SZT
Product Details & Documents

Microsorb™ C18 is an octadecylsilane-covered silica that has been bonded using monofunctional bonding agents under optimised conditions. This material is then endcapped with trimethylsilyl groups to minimise nonspecific binding, maximise mass recoveries and improve peak shape.

C18 columns are typically used with water/methanol or water/acetonitrile solvent combinations and are ideal for the separation of small molecules, proteins, or peptides with differing hydrophobicities.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    100 Å
  • Description:
    Microsorb™ MV
  • Length:
    150 mm
  • Seq:
    02
  • Phase:
    C18