Analytical HPLC columns, Microsorb™ C18
Microsorb™ MV, Analytical, C18, 100 Å, 5 µm, 4,60 mm, 250 mm
Catalog # VARIR0086200C5
Supplier: VARIAN
Jump to:
Product Details & Documents
Microsorb™ C18 is an octadecylsilane-covered silica that has been bonded using monofunctional bonding agents under optimised conditions. This material is then endcapped with trimethylsilyl groups to minimise nonspecific binding, maximise mass recoveries and improve peak shape.
C18 columns are typically used with water/methanol or water/acetonitrile solvent combinations and are ideal for the separation of small molecules, proteins, or peptides with differing hydrophobicities.
Specifications
- Column Format:Analytical
- Ø int.:4,60 mm
- Particle Size:5 µm
- Pore size:100 Å
- Description:Microsorb™ MV
- Length:250 mm
- Seq:03
- Phase:C18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



