Order Entry
Poland
Orders LinkContactUsLinkComponent
Lamivudine, Reference Standards

Lamivudine, Reference Standards

Lamivudine Resolution Mixture A, USP reference standard, Vial, 10 mg
Catalog # 1356847.
Supplier:   USP
Lamivudine, Reference Standards
Catalog # 1356847.
Supplier:  USP
CAS Number:  
Lamivudine, Reference Standards

Lamivudine Resolution Mixture A, USP reference standard, Vial, 10 mg

CAS Number:  
Price
6 750 zł
Log in for availability
1 * 10 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Lamivudine Resolution Mixture B: This is a mixture of lamivudine (about 94%) and lamivudine diastereomer (about 6%)

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    USP reference standard
  • Description:
    Lamivudine Resolution Mixture A

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.