Order Entry
Poland
Orders LinkContactUsLinkComponent
Precision probes, T

Precision probes, T

TC T-type waterproof surface probe with fixed cable, −50…+350 °C, ±0,2 (−20…+70);
±1,0 °C, 5×112 mm
Catalog # 620-3323
Supplier:   TESTO
Precision probes, T
Catalog # 620-3323
Supplier:  TESTO
Precision probes, T
Precision probes, T

TC T-type waterproof surface probe with fixed cable, −50…+350 °C, ±0,2 (−20…+70);
±1,0 °C, 5×112 mm

Price
362 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
Specifications
  • Range:
    −50…+350 °C
  • Description:
    TC T-type waterproof surface probe with fixed cable
  • Seq:
    07
  • Accuracy:
    ±0,2 (−20…+70);
    ±1,0 °C
  • Ø×L:
    5×112 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.