Distilling bridges
200 mm, 19/26 NS
Catalog # WITG1.200.200
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links without condenser, with two vertical NS cones.
Specifications
- Cone size:19/26 NS
- Jacket length:200 mm